Learn More
Analytical Sales & Services Lumidox II 96-Well LED Arrays (Copy)
Catalog No. 5000805162
Encompass
LumiDox Gen2, 96 Well, 470-BLUE LED Array, with Diffuse Mat and Active Cooling Base
Catalog No. 50-008-05162 Supplier Analytical Sales & Services Supplier No. LUM296DA4701
LumiDox Gen2, 96 Well, 470-BLUE LED Array, with Diffuse Mat and Active Cooling Base