Learn More
Analytical Sales & Services Lumidox II 96-Well LED Arrays with Diffuse Mat
Catalog No. 5000805146
Encompass
LumidoxII 96 Well, 505-CYAN LED Array, with Diffuse Mat and Solid Base (requires Thermal Transfer Deck and external chiller)
Catalog No. 50-008-05146 Supplier Analytical Sales & Services Supplier No. CCLUM296DS505
LumidoxII 96 Well, 505-CYAN LED Array, with Diffuse Mat and Solid Base (requires Thermal Transfer Deck and external chiller)