Learn More
Analytical Sales & Services Lumidox II 96-Well LED Arrays with Diffuse Mat
Catalog No. 5000805217
Encompass
LumidoxII 96 Well, 445-INDIGO LED Array, with Diffuse Mat and Solid Base (requires Thermal Transfer Deck and external chiller)
Catalog No. 50-008-05217 Supplier Analytical Sales & Services Supplier No. CCLUM296DS445
LumidoxII 96 Well, 445-INDIGO LED Array, with Diffuse Mat and Solid Base (requires Thermal Transfer Deck and external chiller)