Learn More
Analytical Sales & Services Lumidox II 96-Well LED Arrays with Diffuse Mat
Catalog No. 5000805282
Encompass
LumidoxII 96 Well, 420-VIOLET LED Array, with Diffuse Mat and Flow-through Base (requires external chiller)
Catalog No. 50-008-05282 Supplier Analytical Sales & Services Supplier No. CCLUM296DF420
Add to Cart
Add to Cart
LumidoxII 96 Well, 420-VIOLET LED Array, with Diffuse Mat and Flow-through Base (requires external chiller)