Learn More
Analytical Sales & Services Lumidox II 96-Well LED Arrays with Diffuse Mat
Catalog No. 5000805299
Encompass
LumidoxII 96 Well, 660-DEEP RED LED Array, with Diffuse Mat and Flow-through Base. For deep red, near IR photocatalysis (requires external chiller)
Catalog No. 50-008-05299 Supplier Analytical Sales & Services Supplier No. CCLUM296DF660
LumidoxII 96 Well, 660-DEEP RED LED Array, with Diffuse Mat and Flow-through Base. For deep red, near IR photocatalysis (requires external chiller)