Learn More
Analytical Sales & Services Lumidox II 96-Well LED Arrays with Diffuse Mat
Catalog No. 5000805300
Encompass
LumidoxII 96 Well, UV365 LED Array, with Diffuse Mat and Flow-through Base (requires external chiller)
Catalog No. 50-008-05300 Supplier Analytical Sales & Services Supplier No. CCLUM296DF365
LumidoxII 96 Well, UV365 LED Array, with Diffuse Mat and Flow-through Base (requires external chiller)