Learn More
Analytical Sales & Services Lumidox II 96-Well LED Arrays with Diffuse Mat
Catalog No. 5000805353
Encompass
LumidoxII 96 Well, 730-IR LED Array, with Diffuse Mat and Active Cooling Base. For deep red, near IR photocatalysis.
Catalog No. 50-008-05353 Supplier Analytical Sales & Services Supplier No. CCLUM296DA730
LumidoxII 96 Well, 730-IR LED Array, with Diffuse Mat and Active Cooling Base. For deep red, near IR photocatalysis.