Learn More
Analytical Sales & Services Lumidox II 96-Well LED Arrays with Diffuse Mat
Supplier:  Analytical Sales & Services CCLUM296DS395
Encompass
LumidoxII 96 Well, UV395 LED Array, with Diffuse Mat and Solid Base (requires Thermal Transfer Deck and external chiller)
Catalog No. 50-008-05363