Learn More
Analytical Sales & Services Lumidox II 96-Well LED Arrays with Diffuse Mat
Catalog No. 5000805148
Encompass
LumidoxII 96 Well, 630-RED LED Array, with Diffuse Mat and Solid Base (requires Thermal Transfer Deck and external chiller)
Catalog No. 50-008-05148 Supplier Analytical Sales & Services Supplier No. CCLUM296DS630
Add to Cart
Add to Cart
LumidoxII 96 Well, 630-RED LED Array, with Diffuse Mat and Solid Base (requires Thermal Transfer Deck and external chiller)