Learn More
Analytical Sales & Services Lumidox II 96-Well LED Arrays with Diffuse Mat
Catalog No. 5000807149
Encompass
LumidoxII 96 Well, UV375 LED Array, with Diffuse Mat and Solid Base (requires Thermal Transfer Deck and external chiller)
Catalog No. 50-008-07149 Supplier Analytical Sales & Services Supplier No. CCLUM296DS375
LumidoxII 96 Well, UV375 LED Array, with Diffuse Mat and Solid Base (requires Thermal Transfer Deck and external chiller)