Learn More
Analytical Sales & Services Lumidox II 96-Well LED Arrays with Diffuse Mat
Catalog No. 5000807151
Encompass
LumidoxII 96 Well, 730-IR LED Array, with Diffuse Mat and Solid Base. For deep red, near IR photocatalysis (requires Thermal Transfer Deck and external chiller)
Catalog No. 50-008-07151 Supplier Analytical Sales & Services Supplier No. CCLUM296DS730
LumidoxII 96 Well, 730-IR LED Array, with Diffuse Mat and Solid Base. For deep red, near IR photocatalysis (requires Thermal Transfer Deck and external chiller)