Learn More
Analytical Sales & Services Lumidox II 96-Well LED Arrays with Diffuse Mat
Catalog No. 5000807265
Encompass
LumidoxII 96 Well, 420-VIOLET LED Array, with Diffuse Mat and Solid Base (requires Thermal Transfer Deck and external chiller)
Catalog No. 50-008-07265 Supplier Analytical Sales & Services Supplier No. CCLUM296DS420
LumidoxII 96 Well, 420-VIOLET LED Array, with Diffuse Mat and Solid Base (requires Thermal Transfer Deck and external chiller)