Learn More
Analytical Sales & Services Lumidox II 96-Well LED Arrays with Diffuse Mat
Supplier:  Analytical Sales & Services CCLUM296DF527
Encompass
LumidoxII 96 Well, 527-GREEN LED Array, with Diffuse Mat and Flow-through Base (requires external chiller)
Catalog No. 50-008-05208