Learn More
Analytical Sales & Services Lumidox II 96-Well LED Arrays with Lens Mat
Catalog No. 5000805221
Encompass
LumiDox Gen2, 96 Well, 660-DEEP RED LED Array, with Lens Mat and Active Cooling Base. For deep red, near IR photocatalysis.
Catalog No. 50-008-05221 Supplier Analytical Sales & Services Supplier No. CCLUM296LA660
LumiDox Gen2, 96 Well, 660-DEEP RED LED Array, with Lens Mat and Active Cooling Base. For deep red, near IR photocatalysis.