Learn More
Analytical Sales & Services Lumidox II 96-Well LED Arrays with Lens Mat
Supplier: Analytical Sales & Services CCLUM296LF385
LumiDox Gen2, 96 Well, UV385 LED Array, with Lens Mat and Flow-through Base