Learn More
Analytical Sales & Services Lumidox II 96-Well LED Arrays with Lens Mat
Catalog No. 5000807098
Encompass
LumiDox Gen2, 96 Well, 527-GREEN LED Array, with Lens Mat and Active Cooling Base
Catalog No. 50-008-07098 Supplier Analytical Sales & Services Supplier No. CCLUM296LA527
LumiDox Gen2, 96 Well, 527-GREEN LED Array, with Lens Mat and Active Cooling Base