Learn More
Analytical Sales & Services Lumidox II 96-Well LED Arrays with Lens Mat
Catalog No. 5000807152
Encompass
LumiDox Gen2, 96 Well, 505-CYAN LED Array, with Lens Mat and Active Cooling Base
Catalog No. 50-008-07152 Supplier Analytical Sales & Services Supplier No. CCLUM296LA505
LumiDox Gen2, 96 Well, 505-CYAN LED Array, with Lens Mat and Active Cooling Base