Learn More
Analytical Sales & Services Lumidox II 96-Well LED Arrays with Lens Mat
Catalog No. 5000807197
Encompass
LumiDox Gen2, 96 Well, 420-VIOLET LED Array, with Lens Mat and Solid Base
Catalog No. 50-008-07197 Supplier Analytical Sales & Services Supplier No. CCLUM296LS420
LumiDox Gen2, 96 Well, 420-VIOLET LED Array, with Lens Mat and Solid Base