Learn More
Analytical Sales & Services Lumidox II 96-Well LED Arrays with Lens Mat
Catalog No. 5000807200
Encompass
LumiDox Gen2, 96 Well, UV375 LED Array, with Lens Mat and Solid Base
Catalog No. 50-008-07200 Supplier Analytical Sales & Services Supplier No. CCLUM296LS375
Add to Cart
Add to Cart
LumiDox Gen2, 96 Well, UV375 LED Array, with Lens Mat and Solid Base