Learn More
Analytical Sales & Services Lumidox II 96-Well LED Arrays with Lens Mat and Active Cooling Base
Catalog No. 5000806758
Encompass
LumiDox Gen2, 96 Well, UV365 LED Array, with Lens Mat and Active Cooling Base
Catalog No. 50-008-06758 Supplier Analytical Sales & Services Supplier No. PCRLUM296LA365
LumiDox Gen2, 96 Well, UV365 LED Array, with Lens Mat and Active Cooling Base