Learn More
Analytical Sales & Services Lumidox II LumLamp
Catalog No. 5000805459
Encompass
LumidoxII LumLamp, with 590-AMBER LEDs, 30 Degree Extra Wide Beam Reflector and Clear Polycarbonate Lens
Catalog No. 50-008-05459 Supplier Analytical Sales & Services Supplier No. LUM2L590C30
LumidoxII LumLamp, with 590-AMBER LEDs, 30 Degree Extra Wide Beam Reflector and Clear Polycarbonate Lens