Learn More
Analytical Sales & Services Lumidox II LumLamp
Catalog No. 5000805486
Encompass
LumidoxII LumLamp, with UV405 LEDs, 80 Degree Extra Wide Beam Reflector and Quartz Lens
Catalog No. 50-008-05486 Supplier Analytical Sales & Services Supplier No. LUM2L405Q80
LumidoxII LumLamp, with UV405 LEDs, 80 Degree Extra Wide Beam Reflector and Quartz Lens