Learn More
Analytical Sales & Services Lumidox II LumLamp
Supplier:  Analytical Sales & Services LUM2L505C15
Encompass
LumidoxII LumLamp, with 505-CYAN LEDs, 15 Degree Wide Beam Reflector and Clear Polycarbonate Lens
Catalog No. 50-008-05488