Learn More
Analytical Sales & Services Lumidox II LumLamp
Catalog No. 5000805500
Encompass
LumidoxII LumLamp, with UV385 LEDs, 30 Degree Medium Beam Reflector and Quartz Lens
Catalog No. 50-008-05500 Supplier Analytical Sales & Services Supplier No. LUM2L385Q30
Add to Cart
Add to Cart
LumidoxII LumLamp, with UV385 LEDs, 30 Degree Medium Beam Reflector and Quartz Lens