Learn More
Analytical Sales & Services Lumidox II LumLamp
Catalog No. 5000805501
Encompass
LumidoxII LumLamp, with UV405 LEDs, 30 Degree Medium Beam Reflector and Quartz Lens
Catalog No. 50-008-05501 Supplier Analytical Sales & Services Supplier No. LUM2L405Q30
Add to Cart
Add to Cart
LumidoxII LumLamp, with UV405 LEDs, 30 Degree Medium Beam Reflector and Quartz Lens