Learn More
Analytical Sales & Services Lumidox II LumLamp
Catalog No. 5000805503
Encompass
LumidoxII LumLamp, with 590-AMBER LEDs, 15 Degree Extra Wide Beam Reflector and Diffused Polycarbonate Lens
Catalog No. 50-008-05503 Supplier Analytical Sales & Services Supplier No. LUM2L590D15
LumidoxII LumLamp, with 590-AMBER LEDs, 15 Degree Extra Wide Beam Reflector and Diffused Polycarbonate Lens