Learn More
Analytical Sales & Services Lumidox II LumLamp
Catalog No. 5000805512
Encompass
LumidoxII LumLamp, with 590-AMBER LEDs, 40 Degree Extra Wide Beam Reflector and Quartz Lens
Catalog No. 50-008-05512 Supplier Analytical Sales & Services Supplier No. LUM2L590Q40
Add to Cart
Add to Cart
LumidoxII LumLamp, with 590-AMBER LEDs, 40 Degree Extra Wide Beam Reflector and Quartz Lens