Learn More
Analytical Sales & Services Lumidox II LumLamp
Catalog No. 5000805529
Encompass
LumidoxII LumLamp, with 630-RED LEDs, 30 Degree Extra Wide Beam Reflector and Quartz Lens
Catalog No. 50-008-05529 Supplier Analytical Sales & Services Supplier No. LUM2L630Q30
LumidoxII LumLamp, with 630-RED LEDs, 30 Degree Extra Wide Beam Reflector and Quartz Lens