Learn More
Analytical Sales & Services Lumidox II LumLamp
Catalog No. 5000806165
Encompass
LumidoxII LumLamp, with 445-INDIGO LEDs, 15 Degree Narrow Spot Beam Reflector and Clear Polycarbonate Lens
Catalog No. 50-008-06165 Supplier Analytical Sales & Services Supplier No. LUM2L445C15
LumidoxII LumLamp, with 445-INDIGO LEDs, 15 Degree Narrow Spot Beam Reflector and Clear Polycarbonate Lens