Learn More
Analytical Sales & Services Lumidox II LumLamp
Catalog No. 5000806166
Encompass
LumidoxII LumLamp, with 505-CYAN LEDs, 40 Degree Wide Beam Reflector and Quartz Lens
Catalog No. 50-008-06166 Supplier Analytical Sales & Services Supplier No. LUM2L505Q40
LumidoxII LumLamp, with 505-CYAN LEDs, 40 Degree Wide Beam Reflector and Quartz Lens