Learn More
Analytical Sales & Services Lumidox II LumLamp
Catalog No. 5000806169
Encompass
LumidoxII LumLamp, with 630-RED LEDs, 80 Degree Extra Wide Beam Reflector and Diffused Polycarbonate Lens
Catalog No. 50-008-06169 Supplier Analytical Sales & Services Supplier No. LUM2L630D80
Add to Cart
Add to Cart
LumidoxII LumLamp, with 630-RED LEDs, 80 Degree Extra Wide Beam Reflector and Diffused Polycarbonate Lens