Learn More
Analytical Sales & Services Lumidox II LumLamp
Catalog No. 5000806625
Encompass
LumidoxII LumLamp, with 470-BLUE LEDs, 15 Degree Wide Beam Reflector and Diffused Polycarbonate Lens
Catalog No. 50-008-06625 Supplier Analytical Sales & Services Supplier No. LUM2L470D15
LumidoxII LumLamp, with 470-BLUE LEDs, 15 Degree Wide Beam Reflector and Diffused Polycarbonate Lens