Learn More
Analytical Sales & Services Lumidox II LumLamp
Catalog No. 5000806697
Encompass
LumidoxII LumLamp, with 470-BLUE LEDs, 30 Degree Wide Beam Reflector and Clear Polycarbonate Lens
Catalog No. 50-008-06697 Supplier Analytical Sales & Services Supplier No. LUM2L470C30
LumidoxII LumLamp, with 470-BLUE LEDs, 30 Degree Wide Beam Reflector and Clear Polycarbonate Lens