Learn More
Analytical Sales & Services Lumidox II LumLamp
Catalog No. 5000806726
Encompass
LumidoxII LumLamp, with UV365 LEDs, 30 Degree Medium Beam Reflector and Quartz Lens
Catalog No. 50-008-06726 Supplier Analytical Sales & Services Supplier No. LUM2L365Q30
LumidoxII LumLamp, with UV365 LEDs, 30 Degree Medium Beam Reflector and Quartz Lens