Learn More
Analytical Sales & Services Lumidox II LumLamp
Catalog No. 5000806733
Encompass
LumidoxII LumLamp, with 590-AMBER LEDs, 80 Degree Extra Wide Beam Reflector and Clear Polycarbonate Lens
Catalog No. 50-008-06733 Supplier Analytical Sales & Services Supplier No. LUM2L590C80
LumidoxII LumLamp, with 590-AMBER LEDs, 80 Degree Extra Wide Beam Reflector and Clear Polycarbonate Lens