Learn More
Analytical Sales & Services Lumidox II LumLamp
Catalog No. 5000806735
Encompass
LumidoxII LumLamp, with WHITE LEDs, 30 Degree Extra Wide Beam Reflector and Quartz Lens
Catalog No. 50-008-06735 Supplier Analytical Sales & Services Supplier No. LUM2LWHTQ30
LumidoxII LumLamp, with WHITE LEDs, 30 Degree Extra Wide Beam Reflector and Quartz Lens