Learn More
Analytical Sales & Services Lumidox II LumLamp
Catalog No. 5000806806
Encompass
LumidoxII LumLamp, with UV385 LEDs, 80 Degree Extra Wide Beam Reflector and Quartz Lens
Catalog No. 50-008-06806 Supplier Analytical Sales & Services Supplier No. LUM2L385Q80
LumidoxII LumLamp, with UV385 LEDs, 80 Degree Extra Wide Beam Reflector and Quartz Lens