Learn More
Analytical Sales & Services Lumidox II LumLamp
Catalog No. 5000806807
Encompass
LumidoxII LumLamp, with 505-CYAN LEDs, 15 Degree Wide Beam Reflector and Quartz Lens
Catalog No. 50-008-06807 Supplier Analytical Sales & Services Supplier No. LUM2L505Q15
LumidoxII LumLamp, with 505-CYAN LEDs, 15 Degree Wide Beam Reflector and Quartz Lens