Learn More
Analytical Sales & Services Lumidox II LumLamp
Catalog No. 5000806836
Encompass
LumidoxII LumLamp, with 630-RED LEDs, 15 Degree Extra Wide Beam Reflector and Quartz Lens
Catalog No. 50-008-06836 Supplier Analytical Sales & Services Supplier No. LUM2L630Q15
LumidoxII LumLamp, with 630-RED LEDs, 15 Degree Extra Wide Beam Reflector and Quartz Lens