Learn More
Analytical Sales & Services Lumidox II LumLamp
Catalog No. 5000805455
Encompass
LumidoxII LumLamp, with 505-CYAN LEDs, 80 Degree Extra Wide Beam Reflector and Clear Polycarbonate Lens
Catalog No. 50-008-05455 Supplier Analytical Sales & Services Supplier No. LUM2L505C80
Add to Cart
Add to Cart
LumidoxII LumLamp, with 505-CYAN LEDs, 80 Degree Extra Wide Beam Reflector and Clear Polycarbonate Lens