Learn More
Analytical Sales & Services Lumidox II LumLamp
Catalog No. 5000805456
Encompass
LumidoxII LumLamp, with 470-BLUE LEDs, 30 Degree Medium Beam Reflector and Quartz Lens
Catalog No. 50-008-05456 Supplier Analytical Sales & Services Supplier No. LUM2L470Q30
LumidoxII LumLamp, with 470-BLUE LEDs, 30 Degree Medium Beam Reflector and Quartz Lens