Learn More
Analytical Sales & Services Lumidox II LumLamp
Catalog No. 5000805457
Encompass
LumidoxII LumLamp, with 527-GREEN LEDs, 40 Degree Extra Wide Beam Reflector and Clear Polycarbonate Lens
Catalog No. 50-008-05457 Supplier Analytical Sales & Services Supplier No. LUM2L527C40
LumidoxII LumLamp, with 527-GREEN LEDs, 40 Degree Extra Wide Beam Reflector and Clear Polycarbonate Lens