Learn More
Analytical Sales & Services Lumidox II LumLamp
Catalog No. 5000806858
Encompass
LumidoxII LumLamp, with UV375 LEDs, 40 Degree Wide Beam Reflector and Quartz Lens
Catalog No. 50-008-06858 Supplier Analytical Sales & Services Supplier No. LUM2L375Q40
LumidoxII LumLamp, with UV375 LEDs, 40 Degree Wide Beam Reflector and Quartz Lens