Learn More
Analytical Sales & Services Lumidox II LumLamp
Catalog No. 5000806860
Encompass
LumidoxII LumLamp, with UV405 LEDs, 15 Degree Narrow Spot Beam Reflector and Quartz Lens
Catalog No. 50-008-06860 Supplier Analytical Sales & Services Supplier No. LUM2L405Q15
LumidoxII LumLamp, with UV405 LEDs, 15 Degree Narrow Spot Beam Reflector and Quartz Lens