Learn More
Analytical Sales & Services Lumidox II LumLamp
Catalog No. 5000806863
Encompass
LumidoxII LumLamp, with UV395 LEDs, 40 Degree Wide Beam Reflector and Quartz Lens
Catalog No. 50-008-06863 Supplier Analytical Sales & Services Supplier No. LUM2L395Q40
LumidoxII LumLamp, with UV395 LEDs, 40 Degree Wide Beam Reflector and Quartz Lens