Learn More
Analytical Sales & Services Lumidox II LumLamp
Catalog No. 5000806865
Encompass
LumidoxII LumLamp, with 527-GREEN LEDs, 80 Degree Extra Wide Beam Reflector and Quartz Lens
Catalog No. 50-008-06865 Supplier Analytical Sales & Services Supplier No. LUM2L527Q80
LumidoxII LumLamp, with 527-GREEN LEDs, 80 Degree Extra Wide Beam Reflector and Quartz Lens