Learn More
Analytical Sales & Services Lumidox II LumLamp
Catalog No. 5000807123
Encompass
LumidoxII LumLamp, with 445-INDIGO LEDs, 30 Degree Medium Beam Reflector and Diffused Polycarbonate Lens
Catalog No. 50-008-07123 Supplier Analytical Sales & Services Supplier No. LUM2L445D30
LumidoxII LumLamp, with 445-INDIGO LEDs, 30 Degree Medium Beam Reflector and Diffused Polycarbonate Lens