Learn More
Analytical Sales & Services Lumidox II LumLamp
Catalog No. 5000807125
Encompass
LumidoxII LumLamp, with 445-INDIGO LEDs, 15 Degree Narrow Spot Beam Reflector and Diffused Polycarbonate Lens
Catalog No. 50-008-07125 Supplier Analytical Sales & Services Supplier No. LUM2L445D15
LumidoxII LumLamp, with 445-INDIGO LEDs, 15 Degree Narrow Spot Beam Reflector and Diffused Polycarbonate Lens