Learn More
Analytical Sales & Services Lumidox II LumLamp
Catalog No. 5000807126
Encompass
LumidoxII LumLamp, with 505-CYAN LEDs, 80 Degree Extra Wide Beam Reflector and Quartz Lens
Catalog No. 50-008-07126 Supplier Analytical Sales & Services Supplier No. LUM2L505Q80
LumidoxII LumLamp, with 505-CYAN LEDs, 80 Degree Extra Wide Beam Reflector and Quartz Lens