Learn More
Analytical Sales & Services Lumidox II LumLamp
Catalog No. 5000807128
Encompass
LumidoxII LumLamp, with 470-BLUE LEDs, 80 Degree Extra Wide Beam Reflector and Diffused Polycarbonate Lens
Catalog No. 50-008-07128 Supplier Analytical Sales & Services Supplier No. LUM2L470D80
LumidoxII LumLamp, with 470-BLUE LEDs, 80 Degree Extra Wide Beam Reflector and Diffused Polycarbonate Lens